Learn More
Abnova™ Human HSD17B7 Partial ORF (NP_057455.1, 255 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051478-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The 17-beta-hydroxysteroid dehydrogenase enzyme (EC 1.1.1.62) oxidizes or reduces estrogens and androgens in mammals and regulates the biologic potency of these steroids.[supplied by OMIM]
Sequence: NAFTLTPYNGTEALVWLFHQKPESLNPLIKYLSATTGFGRNYIMTQKMDLDEDTAEKFYQKLLELEKHIRVTIQKTDNQARLSGSCLSpecifications
NP_057455.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NAFTLTPYNGTEALVWLFHQKPESLNPLIKYLSATTGFGRNYIMTQKMDLDEDTAEKFYQKLLELEKHIRVTIQKTDNQARLSGSCL | |
RUO | |
HSD17B7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51478 | |
HSD17B7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12523/MGC75018/PRAP/SDR37C1 | |
HSD17B7 | |
Recombinant | |
wheat germ expression system |