missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSF2BP Full-length ORF (NP_008962.1, 1 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011077-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136142

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation. [provided by RefSeq]

Sequence: MGEAGAAEEACRHMGTKEEFVKVRKKDLERLTTEVMQIRDFLPRILNGEVLESFQKLKIVEKNLERKEQELEQLKMDCEHFKARLETVQADNIREKKEKLALRQQLNEAKQQLLQQAEYCTEMGAAACTLLWGVSSSEEVVKAILGGDKALKFFSITGQTMESFVKSLDGDVQELDSDESQFVFALAGIVTNVAAIACGREFLVNSSRVLLDTILQLLGDLKPGQCTKLKVLMLMSLYNVSINLKGLKYISESPGFIPLLWWLLSDPDAEVCLHVLRLVQSVVLEPEVFSKSASEFRSSLPLQRILAMSKSRNPRLQTAAQELLEDLRTLEHNV
Specifications

Specifications

NP_008962.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSF2BP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
11077
HSF2BP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGEAGAAEEACRHMGTKEEFVKVRKKDLERLTTEVMQIRDFLPRILNGEVLESFQKLKIVEKNLERKEQELEQLKMDCEHFKARLETVQADNIREKKEKLALRQQLNEAKQQLLQQAEYCTEMGAAACTLLWGVSSSEEVVKAILGGDKALKFFSITGQTMESFVKSLDGDVQELDSDESQFVFALAGIVTNVAAIACGREFLVNSSRVLLDTILQLLGDLKPGQCTKLKVLMLMSLYNVSINLKGLKYISESPGFIPLLWWLLSDPDAEVCLHVLRLVQSVVLEPEVFSKSASEFRSSLPLQRILAMSKSRNPRLQTAAQELLEDLRTLEHNV
RUO
HSF2BP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers