Learn More
Invitrogen™ eBioscience™ Human HSP27 Platinum ELISA Kit
For the quantitative detection of human HSP27.
Brand:  Invitrogen™ BMS2086TWO
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15571607
  • 6920.00 SEK / Pack of 2
Estimated Shipment
11-12-2020
Add to basket
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
Biotin-Conjugate anti-human HSP27 monoclonal antibody
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
10 μL
Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1
Human
10.9pg/mL
hsp27
2 x 96 Tests
Serum