Learn More
Invitrogen™ HSP70 Human ELISA Kit
Sandwich ELISA Kit
Brand:  Invitrogen™ BMS2087TEN
27634.71 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15591607
  • 41450.00 SEK / Pack of 10
Estimated Shipment: 10-04-2024
to see stock.
Add to basket
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP70
Biotin-Conjugate anti-human HSP70 monoclonal antibody
Description
Description
The Human Heat Shock Protein 70 (Hu HSP70) ELISA quantitates Hu HSP70 in human serum, plasma, cell lysates or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HSP70. Principle of the method The Human HSP70 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
0.052ng/mL
Human
10 x 96 Tests
HSP70
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
ELISA
Heat Shock Protein 70