missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HSPG2 Partial ORF (NP_005520.3, 25 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005520.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3339 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136471
|
Abnova™
H00003339-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16146471
|
Abnova™
H00003339-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Heparan sulfate proteoglycan is a major component of basement membranes, where the molecule may be involved in the stabilization of other molecules as well as being involved with glomerular permeability to macromolecules and cell adhesion. This form of HSPG, known as HSPG2 or perlecan, is encoded by a gene that maps to chromosome 1. The gene for the form of HSPG associated with the cell surface of fibroblasts has been mapped to human chromosome 8 (MIM 142460).[supplied by OMIM]
Sequence: GLRAYDGLSLPEDIETVTASQMRWTHSYLSDDEDMLADSISGDDLGSGDLGSGDFQMVYFRALVNFTRSIEYSPQLEDAGSREFREVSEAVVDTLESEYLKIPGDQVVSVSpecifications
NP_005520.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLC/PRCAN/SJA/SJS/SJS1 | |
HSPG2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3339 | |
HSPG2 (Human) Recombinant Protein (Q01) | |
GLRAYDGLSLPEDIETVTASQMRWTHSYLSDDEDMLADSISGDDLGSGDLGSGDFQMVYFRALVNFTRSIEYSPQLEDAGSREFREVSEAVVDTLESEYLKIPGDQVVSV | |
RUO | |
HSPG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |