missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HYKK Full-length ORF (ENSP00000353710, 1 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00123688-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117563

  • 6150.00 SEK / 25µg
Estimated Shipment: 04-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSSGNYQQSEALSKPTFSEEQASALVESVFGLKVSKVRPLPSYDDQNFHVYVSKTKDGPTEYVLKISNTKASKNPDLIEVQNHIIMFLKAAGFPTASVCHTKGDNTASLVSVDSGSEIKSYLVRLLTYLPGRPIAELPVSPQLLYEIGKLAAKLDKTLQEGKPRVTPLLAKN
Specifications

Specifications

ENSP00000353710
Liquid
123688
HYKK (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSGNYQQSEALSKPTFSEEQASALVESVFGLKVSKVRPLPSYDDQNFHVYVSKTKDGPTEYVLKISNTKASKNPDLIEVQNHIIMFLKAAGFPTASVCHTKGDNTASLVSVDSGSEIKSYLVRLLTYLPGRPIAELPVSPQLLYEIGKLAAKLDKTLQEGKPRVTPLLAKN
RUO
HYKK
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGPHD1
HYKK
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers