missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IBRDC1 Partial ORF (NP_689766, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00154214-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KVQLGQVEIKCPITECFEFLEETTVVYNLTHEDSIKYKYFLELGRIDSSTKPCPQCKHFTTFKKKGHIPTPSRSESKYKIQCPTCQFVWCFKCHSPWHESpecifications
NP_689766 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KVQLGQVEIKCPITECFEFLEETTVVYNLTHEDSIKYKYFLELGRIDSSTKPCPQCKHFTTFKKKGHIPTPSRSESKYKIQCPTCQFVWCFKCHSPWHE | |
RUO | |
RNF217 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
154214 | |
IBRDC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf172/IBRDC1/MGC26996/dJ84N20.1 | |
RNF217 | |
Recombinant | |
wheat germ expression system |