Learn More
Abnova™ Human IFNK Full-length ORF (Q9P0W0, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056832-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster. [provided by RefSeq]
Sequence: MSTKPDMIQKCLWLEILMGIFIAGTLSLDCNLLNVHLRRVTWQNLRHLSSMSNSFPVECLRENIAFELPQEFLQYTQPMKRDIKKAFYEMSLQAFNIFSQHTFKYWKERHLKQIQIGLDQQAEYLNQCLEEDENENEDMKEMKENEMKPSEARVPQLSSLELRRYFHRIDNFLKEKKYSDCAWEIVRVEIRRCLYYFYKFTALFRRKSpecifications
Q9P0W0 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RP11-27J8.1 | |
IFNK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56832 | |
IFNK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTKPDMIQKCLWLEILMGIFIAGTLSLDCNLLNVHLRRVTWQNLRHLSSMSNSFPVECLRENIAFELPQEFLQYTQPMKRDIKKAFYEMSLQAFNIFSQHTFKYWKERHLKQIQIGLDQQAEYLNQCLEEDENENEDMKEMKENEMKPSEARVPQLSSLELRRYFHRIDNFLKEKKYSDCAWEIVRVEIRRCLYYFYKFTALFRRK | |
RUO | |
IFNK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |