Läs mer
Invitrogen™ Human IgG Monoclonal Antibody (KT131)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549527
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This product was used in an Indirect ELISA. This product does not cross-react with IgA, IgE and IgM.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifikationer
| Human IgG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| KT131 | |
| Unconjugated | |
| Immunoglobulin G; ImmunoglobulinG | |
| Recombinant Human IgG. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |