Learn More
Abnova™ Human IKBKG Partial ORF (NP_003630, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008517-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the regulatory subunit of the inhibitor of kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome. [supplied by RefSeq
Sequence: MNRHLWKSQLCEMVQPSGGPAADQDVLGEESPLGKPAMLHLPSEQGAPETLQRCLEENQELRDAIRQSNQILRERCEELLHFQASQREEKEFLMCKFQEARKLVERLGLESpecifications
NP_003630 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNRHLWKSQLCEMVQPSGGPAADQDVLGEESPLGKPAMLHLPSEQGAPETLQRCLEENQELRDAIRQSNQILRERCEELLHFQASQREEKEFLMCKFQEARKLVERLGLE | |
RUO | |
IKBKG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8517 | |
IKBKG (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMCBX1/FIP-3/FIP3/Fip3p/IKK-gamma/IP/IP1/IP2/IPD2/NEMO | |
IKBKG | |
Recombinant | |
wheat germ expression system |