Läs mer
R&D Systems™ Human IL-11 R alpha, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8895-MR-050
Beskrivning
The Recombinant Human IL-11 R alpha Protein is derived from CHO. The Recombinant Human IL-11 R alpha Protein has been validated for the following applications: Bioactivity.Specifikationer
| Q14626 | |
| Observed M.W.: 40-55kDa, reducing conditions, Theoretical M.W.: 38kDa | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1 | |
| IL11RA | |
| Unconjugated |
| 3590 | |
| IL-11R alpha | |
| Chinese Hamster Ovary cell line,CHO-derived human IL-11 R alpha protein Ser24-Val363,with a C-terminal 6-His tag | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| IL-11 R alpha | |
| Measured by its ability to bind recombinant human IL-11 in a functional ELISA with an estimated Kd <1 nM. | |
| Yes |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.