missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human IL-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB213-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL-13 Monoclonal specifically detects IL-13 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifikationer
| IL-13 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.15-0.75 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P35225 | |
| IL13 | |
| E. coli-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225 | |
| 100 μg | |
| Primary | |
| Detects human IL-13 in direct ELISAs. | |
| Human | |
| Supernatant |
| Flow Cytometry, Neutralization, CyTOF, ELISA | |
| 32116 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3596 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering