missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human IL-16 C-terminal Peptide Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems BAF316
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL-16 Polyclonal specifically detects IL-16 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, ELISA Detection (Matched Antibody Pair).
Specifikationer
| IL-16 | |
| Polyclonal | |
| Biotin | |
| Q14005 | |
| IL16 | |
| E. coli-derived recombinant human IL-16 isoform 1 Met1203-Ser1332 Accession # Q14005 | |
| 50 μg | |
| Primary | |
| Detects human IL-16 in ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse IL-16 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3603 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering