missing translation for 'onlineSavingsMsg'
Läs mer

Gibco™ Human IL-19, Animal-Free Recombinant Protein, PeproTech®

Product Code. 30019710
Click to view available options
Kvantitet:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
This item is not returnable. View return policy

Product Code. 30019710

Brand: Gibco™ AF20019250UG

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-19 is a 35.8 kDa homodimer of two 154 amino acid chains. In solution, IL-19 exists predominantly as a non-disulfide-linked dimer.

The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Tillträdesnummer Q9UHD0
För användning med (applikation) Functional Assay
Formatera Lyophilized
Formulering protein with no preservative
Gen-ID (Entrez) 29949
Molekylvikt (g/mol) 35.8 kDa
Namn Human IL-19, Animal-Free
Kvantitet 250 μg
Regulatorisk status RUO
Endotoxinkoncentration <1 EU/ μg
Gene Alias il 19; IL-10 C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG0.1; ZMDA1
Vanligt namn IL-19
Biologisk aktivitet Determined by its ability to stimulate the proliferation of HEK-Blue TM-IL-19 reporter cells
Konjugera Unconjugated
Rekombinant Recombinant
Sekvens MLRRCLISTD MHHIEESFQE IKRAIQAKDT FPNVTILSTL ETLQIIKPLD VCCVTKNLLA FYVDRVFKDH QEPNPKILRK ISSIANSFLY MQKTLRQCQE QRQCHCRQEA TNATRVIHDN YDQLEVHAAA IKSLGELDVF LAWINKNHEV MSSA
Innehåll och lagring -20°C
Uttryckssystem E. coli
Form Lyophilized
Renhet eller kvalitetsklass ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.