missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Human IL-27 R alpha/WSX-1/TCCR (NP_004834) VersaClone cDNA

Produktkod. 18778849
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18778849 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18778849 Leverantör R&D Systems™ Leverantörsnummer RDC1225

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

IL27RA is involved in the regulation of Th1 type immune responses. It may also be involved in innate defense mechanisms

The Human IL-27 R alpha/WSX-1/TCCR (NP_004834) VersaClone cDNA clone contains the complete ORF for IL-27R alpha/WSX-1/TCCR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-27R alpha/WSX-1/TCCR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer NP_004834
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.5
Infoga typ 1924bp
Gene Alias class I cytokine receptor, CRL1IL-27 R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27 RA, IL-27 R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WS x 1 IL-27 R subunit alpha, zcytor1
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Human
Kvantitet 10 μg

For Reasearch Use Only; Not for Use in Humans

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.