Läs mer
R&D Systems™ Human IL-36 beta/IL-1F8 (NP_775270) VersaClone cDNA
Produktkod. 18720769
Change view
Kvantitet:
10 μg
Produktkod. Kvantitet
18720769 10 μg
1 options
Produktkod. 18720769 Leverantör R&D Systems™ Leverantörsnummer RDC2447
  • ONLINE EXCLUSIVE PRICE
    3500.00 SEK
    3309.60 SEK / 10µg
    Save 190.40 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 19-10-2026
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Human IL-36 beta/IL-1F8 (NP_775270) VersaClone cDNA clone contains the complete ORF for IL-36 beta/IL-1F8, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IL-36 beta/IL-1F8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifikationer
Gen hIL-36 beta/IL-1F8
Tillträdesnummer NP_775270
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-ID (Entrez) 27177
Innehåll och lagring Store the unopened product at -20 to -80°C
Art Human
Kvantitet 10 μg