missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Activin A, Recombinant Protein, Carrier-Free

Human Recombinant Protein

Brand:  Invitrogen™ 34-8993-82

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15518846

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Description

Description

Activin A is a member of the TGF-β superfamily, along with the other activin proteins and the structurally related inhibins. The activins are dimeric proteins formed by the association of two of the four existing β subunits, βA, βB, βC, and βE. These subunits are shared with the inhibin proteins, which are heterodimers of one β and one α chain. Activin A is a homodimer of two 13 kDa βA subunits, which can also associate with βB or βC to form Activin AB or AC, respectively. Activin A is expressed by various cell types and exhibits pleiotropic effects including the regulation of metabolism, homeostasis, cell proliferation and differentiation, apoptosis, and tissue healing. Activin A has been recently reported to work in synergy with TGF-β to promote the induction of antigen specific iTreg cells leading to inhibition of Th1 and Th2-mediated responses.
Specifications

Specifications

NM_002192
Cytokine Bioassays
26kDa
SDS-PAGE
100μg
Store at less than or equal to -70°C.
Insect expressed amino acids Gly311-Ser426, accession number NM_002192
The ED50 of this protein, as measured by inhibition of MPC-11 mouse plasmacytoma cell proliferation, is 0.75-3ng/mL. This corresponds to a specific activity of 1.3 x 106 - 3.3 x 105 Units/mg.
Unlabeled
Yes
0.5mg/mL
Sterile liquid: 20mM glycine, 150mM NaCl pH 3.0. 0.22μm filtered.
Activin A
≥95%
Insect Cells
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
Recombinant Protein, Carrier-Free
Human
Videos