missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IL27 Full-length ORF (AAH62422.1, 1 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00246778-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179573

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR). [provided by RefSeq]

Sequence: MGQTAGDLGWRLSLLLLPLLLVQAGVWGFPRPPGRPQLSLQELRREFTVSLHLARKLLAEVRGQAHRFAESHLPGVNLYLLPLGEQLPDVSLTFQAWRRLSDPERLCFISTTLQPFHALLGGLGTQGRWTNMERMQLWAMRLDLRDLQRHLRFQVLAAGFNLPEEEEEEEEEEEEERKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQP
Specifications

Specifications

AAH62422.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IL-27/IL-27A/IL27p28/IL30/MGC71873/p28
IL27
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
246778
IL27 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGQTAGDLGWRLSLLLLPLLLVQAGVWGFPRPPGRPQLSLQELRREFTVSLHLARKLLAEVRGQAHRFAESHLPGVNLYLLPLGEQLPDVSLTFQAWRRLSDPERLCFISTTLQPFHALLGGLGTQGRWTNMERMQLWAMRLDLRDLQRHLRFQVLAAGFNLPEEEEEEEEEEEEERKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQP
RUO
IL27
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers