Learn More
Abnova™ Human IL27 Partial ORF (NP_663634, 177 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00246778-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR). [provided by RefSeq]
Sequence: RKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQPSpecifications
NP_663634 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.11kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQP | |
RUO | |
IL27 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
246778 | |
IL27 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL-27/IL-27A/IL27p28/IL30/MGC71873/p28 | |
IL27 | |
Recombinant | |
wheat germ expression system |