missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human INTS4 Full-length ORF (NP_291025.3, 1 a.a. - 963 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_291025.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 92105 |
Molecular Weight (g/mol) | 134.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136693
|
Abnova™
H00092105-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126693
|
Abnova™
H00092105-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM]
Sequence: MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARRSpecifications
NP_291025.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
134.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARR | |
RUO | |
INTS4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
92105 | |
INTS4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
INT4/MGC16733/MST093 | |
INTS4 | |
Recombinant | |
wheat germ expression system |