missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human INTS9 Full-length ORF (AAH25267.1, 1 a.a. - 658 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055756-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16171763

  • 6845.00 SEK / 25µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM]

Sequence: MKLYCLSGHPTLPCNVLKFKSTTIMLDCGLDMTSTLNFLPLPLVQSPRLSNLPGWSLKDGNAFLDKELKECSGHVFVDSVPEFCLPETELIDLSTVDVILISNYHCMMALPYITEHTGFTGTVYATEPTVQIGRLLMEELVNFIERVPKAQSASLWKNKDIQRLLPSPLKDAVEVSTWRRCYTMQEVNSALSKIQLVGYSQKIELFGAVQVTPLSSGYALGSSNWIIQSHYEKVSYVSGSSLLTTHPQPMDQASLKNSDVLVLTGLTQIPTANPDGMVGEFCSNLALTVRNGGNVLVPCYPSGVIYDLLECLYQYIDSAGLSSVPLYFISPVANSSLEFSQIFAEWLCHNKQSKVYLPEPPFPHAELIQTNKLKHYPSIHGDFSNDFRQPCVVFTGHPSLRFGDVVHFMELWGKSSLNTVIFTEPDFSYLEALAPYQPLAMKCIYCPIDTRLNFIQVSKLLKEVQPLHVVCPEQYTQPPPAQSHRMDLMIDCQPPAMSYRRAEVLALPFKRRYEKIEIMPELADSLVPMEIKPGISLATVSAVLHTKDNKHLLQPPPRPAQPTSGKKRKRVSDDVPDCKVLKPLLSGSIPVEQFVQTLEKHGFSDIKVEDTAKGHIVLLQEAETLIQIEEDSTHIICDNDEMLRVRLRDLVLKFLQKF
Specifications

Specifications

AAH25267.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CPSF2L/FLJ10871/INT9/RC-74/RC74
INTS9
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55756
INTS9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLYCLSGHPTLPCNVLKFKSTTIMLDCGLDMTSTLNFLPLPLVQSPRLSNLPGWSLKDGNAFLDKELKECSGHVFVDSVPEFCLPETELIDLSTVDVILISNYHCMMALPYITEHTGFTGTVYATEPTVQIGRLLMEELVNFIERVPKAQSASLWKNKDIQRLLPSPLKDAVEVSTWRRCYTMQEVNSALSKIQLVGYSQKIELFGAVQVTPLSSGYALGSSNWIIQSHYEKVSYVSGSSLLTTHPQPMDQASLKNSDVLVLTGLTQIPTANPDGMVGEFCSNLALTVRNGGNVLVPCYPSGVIYDLLECLYQYIDSAGLSSVPLYFISPVANSSLEFSQIFAEWLCHNKQSKVYLPEPPFPHAELIQTNKLKHYPSIHGDFSNDFRQPCVVFTGHPSLRFGDVVHFMELWGKSSLNTVIFTEPDFSYLEALAPYQPLAMKCIYCPIDTRLNFIQVSKLLKEVQPLHVVCPEQYTQPPPAQSHRMDLMIDCQPPAMSYRRAEVLALPFKRRYEKIEIMPELADSLVPMEIKPGISLATVSAVLHTKDNKHLLQPPPRPAQPTSGKKRKRVSDDVPDCKVLKPLLSGSIPVEQFVQTLEKHGFSDIKVEDTAKGHIVLLQEAETLIQIEEDSTHIICDNDEMLRVRLRDLVLKFLQKF
RUO
INTS9
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers