missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IRGM Partial ORF (XP_293893.3, 99 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | XP_293893.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 345611 |
Molecular Weight (g/mol) | 34.87kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142857
|
Abnova™
H00345611-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16132857
|
Abnova™
H00345611-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the p47 immunity-related GTPase (IRG) family, including IRGM, play an important role in the murine immune system; however, in humans this resistance system is greatly reduced. There is evidence that human IRGM plays a role in autophagy and control of intracellular mycobacteria (Bekpen et al., 2005; Singh et al., 2006 [PubMed 16888103]).[supplied by OMIM]
Sequence: TTLENYLMEMQFNRYDFIMVASAQFSMNHVMLAKTAEDMGKKFYIVWTKLDMDLSTGALPEVQLLQIRENVLENLQKERVCEYSpecifications
XP_293893.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IFI1/IRGM1/LRG-47/LRG47/MGC149263/MGC149264 | |
IRGM | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
345611 | |
IRGM (Human) Recombinant Protein (Q01) | |
TTLENYLMEMQFNRYDFIMVASAQFSMNHVMLAKTAEDMGKKFYIVWTKLDMDLSTGALPEVQLLQIRENVLENLQKERVCEY | |
RUO | |
IRGM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |