missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IVNS1ABP Partial ORF (NP_006460.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010625-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MIPNGYLMFEDENFIESSVAKLNALRKSGQFCDVRLQVCGHEMLAHRAVLACCSPYLFEIFNSDSDPHGISHVKFDDLNPEAVEVLLNYAYTAQLKADKESpecifications
NP_006460.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIPNGYLMFEDENFIESSVAKLNALRKSGQFCDVRLQVCGHEMLAHRAVLACCSPYLFEIFNSDSDPHGISHVKFDDLNPEAVEVLLNYAYTAQLKADKE | |
RUO | |
IVNS1ABP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10625 | |
IVNS1ABP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686K06216/FLARA3/FLJ10069/FLJ10411/FLJ10962/FLJ35593/FLJ36593/HSPC068/KIAA0850/ND1/NS-1/NS1-BP/NS1BP | |
IVNS1ABP | |
Recombinant | |
wheat germ expression system |