Learn More
Abnova™ Human JAG2 Partial ORF (NP_002217.3, 869 a.a. - 966 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003714-Q02.10ug
Additional Details : Weight : 0.00010kg
Description
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. The protein encoded by this gene is one of several ligands that activate Notch and related receptors. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GRSCWSRGTPFPHGSSWVEDCNSCRCLDGRRDCSKVWCGWKPCLLAGQPEALSAQCPLGQRCLEKAPGQCLRPPCEAWGECGAEEPPSTPCLPRSGHLSpecifications
NP_002217.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GRSCWSRGTPFPHGSSWVEDCNSCRCLDGRRDCSKVWCGWKPCLLAGQPEALSAQCPLGQRCLEKAPGQCLRPPCEAWGECGAEEPPSTPCLPRSGHL | |
RUO | |
JAG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3714 | |
JAG2 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HJ2/SER2 | |
JAG2 | |
Recombinant | |
wheat germ expression system |