missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JARID1A Partial ORF (NP_005047, 191 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005927-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194155

  • 6845.00 SEK / 25µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein which regulates cell proliferation. This protein also interacts with rhombotin-2 which functions distinctly in erythropoiesis and in T-cell leukemogenesis. Rhombotin-2 is thought to either directly affect the activity of the encoded protein or may indirectly modulate the functions of the retinoblastoma protein by binding to this protein. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Sequence: KVEPEVLSTDTQTSPEPGTRMNILPKRTRRVKTQSESGDVSRNTELKKLQIFGAGPKVVGLAMGTKDKEDEVTRRRKVTNRSDAFNMQMRQRKGTLSVNF
Specifications

Specifications

NP_005047
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KVEPEVLSTDTQTSPEPGTRMNILPKRTRRVKTQSESGDVSRNTELKKLQIFGAGPKVVGLAMGTKDKEDEVTRRRKVTNRSDAFNMQMRQRKGTLSVNF
RUO
JARID1A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5927
JARID1A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KDM5A/RBBP2/RBP2
JARID1A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers