missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JARID1D Partial ORF (NP_004644.2, 127 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008284-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138365

  • 6845.00 SEK / 25µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein containing zinc finger domains. A short peptide derived from this protein is a minor histocompatibility antigen which can lead to graft rejection of male donor cells in a female recipient. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: AICKDRRWARVAQRLHYPPGKNIGSLLRSHYERIIYPYEMFQSGANHVQCNTHPFDNEVKDKEYKPHSIPLRQSVQPSKFSSYSRRAKRLQPDPEPTEEDIEKH
Specifications

Specifications

NP_004644.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
12.5% SDS-PAGE Stained with Coomassie Blue
AICKDRRWARVAQRLHYPPGKNIGSLLRSHYERIIYPYEMFQSGANHVQCNTHPFDNEVKDKEYKPHSIPLRQSVQPSKFSSYSRRAKRLQPDPEPTEEDIEKH
RUO
JARID1D
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8284
JARID1D (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HY/HYA/KDM5D/KIAA0234/SMCY
JARID1D
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers