Learn More
Abnova™ Human KCNH6 Partial ORF (NP_110406.1, 31 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00081033-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. Several alternatively spliced transcript variants have been identified from this gene, but the full-length nature of only two transcript variants has been determined. [provided by RefSeq]
Sequence: IANAQMENCAIIYCNDGFCELFGYSRVEVMQQPCTCDFLTGPNTPSSAVSRLAQALLGAEECKVDILYYRKDASSFRCLVSpecifications
NP_110406.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IANAQMENCAIIYCNDGFCELFGYSRVEVMQQPCTCDFLTGPNTPSSAVSRLAQALLGAEECKVDILYYRKDASSFRCLV | |
RUO | |
KCNH6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
81033 | |
KCNH6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERG2/HERG2/Kv11.2 | |
KCNH6 | |
Recombinant | |
wheat germ expression system |