missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KCNMB1 Partial ORF (AAH25707, 39 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003779-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137111

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the product of this gene, the modulatory beta subunit. Intracellular calcium regulates the physical association between the alpha and beta subunits. [provided by RefSeq]

Sequence: LPLYQKSVWTQESKCHLIETNIRDQEELKGKKVPQYPCLWVNVSAAGRWAVLYHTEDTRDQNQQVLNWRDGDTSLYPCQVCEPVPNCPCPRG
Specifications

Specifications

AAH25707
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LPLYQKSVWTQESKCHLIETNIRDQEELKGKKVPQYPCLWVNVSAAGRWAVLYHTEDTRDQNQQVLNWRDGDTSLYPCQVCEPVPNCPCPRG
RUO
KCNMB1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3779
KCNMB1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
K(VCA)beta/SLO-BETA/hslo-beta
KCNMB1
Yes
Videos
SDS
Documents
Certificates