missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KCNRG Partial ORF (NP_775876.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_775876.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 283518 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182707
|
Abnova™
H00283518-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172707
|
Abnova™
H00283518-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KCNRG is a soluble protein with characteristics suggesting it forms heterotetramers with voltage-gated K(+) channels (see MIM 176260) and inhibits their function (Ivanov et al., 2003 [PubMed 12650944]).[supplied by OMIM]
Sequence: MSSQELVTLNVGGKIFTTRFSTIKQFPASRLARMLDGRDQEFKMVGGQIFVDRDGDLFSFILDFLRTHQLLLPTEFSDYLRLQREALFYELRSLVDLLNPSpecifications
NP_775876.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLTET | |
KCNRG | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
283518 | |
KCNRG (Human) Recombinant Protein (Q01) | |
MSSQELVTLNVGGKIFTTRFSTIKQFPASRLARMLDGRDQEFKMVGGQIFVDRDGDLFSFILDFLRTHQLLLPTEFSDYLRLQREALFYELRSLVDLLNP | |
RUO | |
KCNRG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |