missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KERA Partial ORF (NP_008966, 253 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011081-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133666

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Keratan sulfate proteoglycans (KSPGs) are members of the small leucine-rich proteoglycan (SLRP) family. KSPGs, particularly keratocan, lumican (MIM 600616), and mimecan (MIM 602383), are important to the transparency of the cornea.[supplied by OMIM]

Sequence: GLPSRGFDVSSILDLQLSHNQLTKVPRISAHLQHLHLDHNKIKSVNVSVICPSPSMLPAERDSFSYGPHLRYLRLDGNEIKPPIPMALMTCFRLLQAVI
Specifications

Specifications

NP_008966
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GLPSRGFDVSSILDLQLSHNQLTKVPRISAHLQHLHLDHNKIKSVNVSVICPSPSMLPAERDSFSYGPHLRYLRLDGNEIKPPIPMALMTCFRLLQAVI
RUO
KERA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11081
KERA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CNA2/SLRR2B
KERA
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers