missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1201 Partial ORF (XP_370660, 250 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057476-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132713

  • 5030.41 SEK / 25µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: NTMGYCEEIPVEENEVNDSSSKSSIETKPDASPQLPKKSITNSTLTSTGSSEAPVSFDGLPLEEEALEGDGSLEKELAIDNIMGEKIEMIAPVNSPSLDFNDNE
Specifications

Specifications

XP_370660
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NTMGYCEEIPVEENEVNDSSSKSSIETKPDASPQLPKKSITNSTLTSTGSSEAPVSFDGLPLEEEALEGDGSLEKELAIDNIMGEKIEMIAPVNSPSLDFNDNE
RUO
GRAMD1B
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
57476
KIAA1201 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1201/MGC125622/MGC125623/MGC125625
GRAMD1B
Yes
Videos
SDS
Documents
Certificates