missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIF15 Partial ORF (NP_064627, 1279 a.a. - 1385 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056992-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122403

  • 6845.00 SEK / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: NKEMECLRMTDEVERTQTLESKAFQEKEQLRSKLEEMYEERERTSQEMEMLRKQVECLAEENGKLVGHQNLHQKIQYVVRLKKENVRLAEETEKLRAENVFLKEKKR
Specifications

Specifications

NP_064627
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NKEMECLRMTDEVERTQTLESKAFQEKEQLRSKLEEMYEERERTSQEMEMLRKQVECLAEENGKLVGHQNLHQKIQYVVRLKKENVRLAEETEKLRAENVFLKEKKR
RUO
KIF15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56992
KIF15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25667/HKLP2/KNSL7/NY-BR-62
KIF15
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers