missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIFC1 Partial ORF (AAH00712, 53 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003833-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117171

  • 3860.70 SEK / 10µg
Estimated Shipment: 27-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MDPQRSPLLEVKGNIELKRPLIKAPSQLPLSGSRLKRRPDQMEDGLEPEKKRTRGLGATTKITTSHPRVPSLTTVPQTQGQTTAQKVSKKTGPRCSTAIA
Specifications

Specifications

AAH00712
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDPQRSPLLEVKGNIELKRPLIKAPSQLPLSGSRLKRRPDQMEDGLEPEKKRTRGLGATTKITTSHPRVPSLTTVPQTQGQTTAQKVSKKTGPRCSTAIA
RUO
KIFC1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3833
KIFC1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSET/KNSL2/MGC1202/MGC149736/MGC149737
KIFC1
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers