Learn More
Abnova™ Human Kir2Ds5 Full-Length Orf (Aai60061.1, 1 A.A.-304 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAI60061.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3810 |
Name | KIR2DS5 (Human) Recombinant Protein (P01) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127141
|
Abnova™
H00003810-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137141
|
Abnova™
H00003810-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MSLMVISMACVAFFLLQGAWPHEGFRRKPSLLAHPGPLVKSEETVILQCWSDVMFEHFLLHREGTFNHTLRLIGEHIDGVSKGNFSIGRMTQDLAGTYRCYGSVTHSPYQLSAPSDPLDIVITGLYEKPSLSAQPGPTVLAGESVT
Specifications
AAI60061.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
KIR2DS5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD158G/NKAT9 | |
KIR2DS5 | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3810 | |
Glutathione Sepharose 4 Fast Flow | |
MSLMVISMACVAFFLLQGAWPHEGFRRKPSLLAHPGPLVKSEETVILQCWSDVMFEHFLLHREGTFNHTLRLIGEHIDGVSKGNFSIGRMTQDLAGTYRCYGSVTHSPYQLSAPSDPLDIVITGLYEKPSLSAQPGPTVLAGESVTLSCSSRSSYDMYHLSREGEAHERRLPAGPKVNRTFQADFPLDPATHGGTYRCFGSFRDSPYEWSKSSDPLLVSVTGNSSNSWPSPTEPSSETGNPRHLHVLIGTSVVKLPFTILLFFLLHRWCSNKKNASVMDQGPAGNRTVNREDSDEQDHQEVSYA | |
RUO | |
KIR2DS5 | |
GST |
For Research Use Only