Learn More
Abnova™ Human KPNA6 Full-length ORF (NP_036448.1, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_036448.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23633 |
Molecular Weight (g/mol) | 86.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107572
|
Abnova™
H00023633-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16117572
|
Abnova™
H00023633-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. The protein encoded by this gene is a member of the importin alpha family. [provided by RefSeq]
Sequence: METMASPGKDNYRMKSYKNNALNPEEMRRRREEEGIQLRKQKREQQLFKRRNVELINEEAAMFDSLLMDSYVSSTTGESVITREMVEMLFSDDSDLQLATTQKFRKLLSKEPSPPIDEVINTPRVVDRFVEFLKRNENCTLQFEAAWALTNIASGTSQQTKIVIEAGAVPIFIELLNSDFEDVQEQAVWALGNIAGDSSVCRDYVLNCSILNPLLTLLTKSTRLTMTRNAVWALSNLCRGKNPPPEFAKVSPCLPVLSRLLFSSDSDLLADACWALSYLSDGPNEKIQAVIDSGVCRRLVELLMHNDYKVASPALRAVGNIVTGDDIQTQVILNCSALPCLLHLLSSPKESIRKEACWTISNITAGNRAQIQAVIDANIFPVLIEILQKAEFRTRKEAAWAITNATSGGTPEQIRYLVSLGCIKPLCDLLTVMDSKIVQVALNGLENILRLGEQEGKRSGSGVNPYCGLIEEAYGLDKIEFLQSHENQEIYQKAFDLIEHYFGVEDDDSSLAPQVDETQQQFIFQQPEAPMEGFQLSpecifications
NP_036448.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
METMASPGKDNYRMKSYKNNALNPEEMRRRREEEGIQLRKQKREQQLFKRRNVELINEEAAMFDSLLMDSYVSSTTGESVITREMVEMLFSDDSDLQLATTQKFRKLLSKEPSPPIDEVINTPRVVDRFVEFLKRNENCTLQFEAAWALTNIASGTSQQTKIVIEAGAVPIFIELLNSDFEDVQEQAVWALGNIAGDSSVCRDYVLNCSILNPLLTLLTKSTRLTMTRNAVWALSNLCRGKNPPPEFAKVSPCLPVLSRLLFSSDSDLLADACWALSYLSDGPNEKIQAVIDSGVCRRLVELLMHNDYKVASPALRAVGNIVTGDDIQTQVILNCSALPCLLHLLSSPKESIRKEACWTISNITAGNRAQIQAVIDANIFPVLIEILQKAEFRTRKEAAWAITNATSGGTPEQIRYLVSLGCIKPLCDLLTVMDSKIVQVALNGLENILRLGEQEGKRSGSGVNPYCGLIEEAYGLDKIEFLQSHENQEIYQKAFDLIEHYFGVEDDDSSLAPQVDETQQQFIFQQPEAPMEGFQL | |
RUO | |
KPNA6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23633 | |
KPNA6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11249/IPOA7/KPNA7/MGC17918 | |
KPNA6 | |
Recombinant | |
wheat germ expression system |