Learn More
Abnova™ Human KRT3 Partial ORF (NP_476429.1, 232 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003850-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the corneal epithelium with family member KRT12 and mutations in these genes have been associated with Meesmann's Corneal Dystrophy. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. [provided by RefSeq]
Sequence: LLQQQGTSSISGTNNLEPLFENHINYLRSYLDNILGERGRLDSELKNMEDLVEDFKKKSpecifications
NP_476429.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.12kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLQQQGTSSISGTNNLEPLFENHINYLRSYLDNILGERGRLDSELKNMEDLVEDFKKK | |
RUO | |
KRT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3850 | |
KRT3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CK3/FLJ95909/K3 | |
KRT3 | |
Recombinant | |
wheat germ expression system |