missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KRT32 Full-length ORF (AAI48741.1, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAI48741.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3882 |
Molecular Weight (g/mol) | 76.23kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127251
|
Abnova™
H00003882-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137251
|
Abnova™
H00003882-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]
Sequence: MTSSCCVTNNLQASLKSCPRPASVCSSGVNCRPELCLGYVCQPMACLPSVCLPTTFRPASCLSKTYLSSSCQAASGISGSMGPGSWYSEGAFNGNEKETMQFLNDRLASYLTRVRQLEQENAELESRIQEASHSQVLTMTPDYQSHFRTIEELQQKILCTKAENARMVVNIDNAKLAADDFRAKYEAELAMRQLVEADINGLRRILDDLTLCKADLEAQVESLKEELMCLKKNHEEEVGSLRCQLGDRLNIEVDAAPPVDLTRVLEEMRCQYEAMVEANRRDVEEWFNMQMEELNQQVATSSEQLQNYQSDIIDLRRTVNTLEIELQAQHSLRDSLENTLTESEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLEGEINTYRSLLENEDCKLPCNPCSTPSCTTCVPSPCVPRTVCVPRTVGMPCSPCPQGRYSpecifications
AAI48741.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.23kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTSSCCVTNNLQASLKSCPRPASVCSSGVNCRPELCLGYVCQPMACLPSVCLPTTFRPASCLSKTYLSSSCQAASGISGSMGPGSWYSEGAFNGNEKETMQFLNDRLASYLTRVRQLEQENAELESRIQEASHSQVLTMTPDYQSHFRTIEELQQKILCTKAENARMVVNIDNAKLAADDFRAKYEAELAMRQLVEADINGLRRILDDLTLCKADLEAQVESLKEELMCLKKNHEEEVGSLRCQLGDRLNIEVDAAPPVDLTRVLEEMRCQYEAMVEANRRDVEEWFNMQMEELNQQVATSSEQLQNYQSDIIDLRRTVNTLEIELQAQHSLRDSLENTLTESEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLEGEINTYRSLLENEDCKLPCNPCSTPSCTTCVPSPCVPRTVCVPRTVGMPCSPCPQGRY | |
RUO | |
KRT32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3882 | |
KRT32 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HA2/HKA2/KRTHA2/hHa2 | |
KRT32 | |
Recombinant | |
wheat germ expression system |