missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT32 Full-length ORF (AAI48741.1, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003882-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137251

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]

Sequence: MTSSCCVTNNLQASLKSCPRPASVCSSGVNCRPELCLGYVCQPMACLPSVCLPTTFRPASCLSKTYLSSSCQAASGISGSMGPGSWYSEGAFNGNEKETMQFLNDRLASYLTRVRQLEQENAELESRIQEASHSQVLTMTPDYQSHFRTIEELQQKILCTKAENARMVVNIDNAKLAADDFRAKYEAELAMRQLVEADINGLRRILDDLTLCKADLEAQVESLKEELMCLKKNHEEEVGSLRCQLGDRLNIEVDAAPPVDLTRVLEEMRCQYEAMVEANRRDVEEWFNMQMEELNQQVATSSEQLQNYQSDIIDLRRTVNTLEIELQAQHSLRDSLENTLTESEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLEGEINTYRSLLENEDCKLPCNPCSTPSCTTCVPSPCVPRTVCVPRTVGMPCSPCPQGRY
Specifications

Specifications

AAI48741.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.23kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HA2/HKA2/KRTHA2/hHa2
KRT32
Yes
wheat germ expression system
Liquid
3882
KRT32 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSSCCVTNNLQASLKSCPRPASVCSSGVNCRPELCLGYVCQPMACLPSVCLPTTFRPASCLSKTYLSSSCQAASGISGSMGPGSWYSEGAFNGNEKETMQFLNDRLASYLTRVRQLEQENAELESRIQEASHSQVLTMTPDYQSHFRTIEELQQKILCTKAENARMVVNIDNAKLAADDFRAKYEAELAMRQLVEADINGLRRILDDLTLCKADLEAQVESLKEELMCLKKNHEEEVGSLRCQLGDRLNIEVDAAPPVDLTRVLEEMRCQYEAMVEANRRDVEEWFNMQMEELNQQVATSSEQLQNYQSDIIDLRRTVNTLEIELQAQHSLRDSLENTLTESEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLEGEINTYRSLLENEDCKLPCNPCSTPSCTTCVPSPCVPRTVCVPRTVGMPCSPCPQGRY
RUO
KRT32
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates