Learn More
Abcam Human KRTCAP2 (KCP-2) knockout HEK-293T cell line
Product Code. 32139816 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32139816 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32139816 Supplier Abcam Supplier No. ab2667192x1000000Cells_
 Restricted Item
Beräknad leverans: 27-11-2026
KRTCAP2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only ...
KRTCAP2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO