missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Human KV3.1 (KCNC1) (aa 116-160) Control Fragment Recombinant Protein
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83873 (PA5-83873. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifikationer
Specifikationer
| Tillträdesnummer | P48547 |
| Koncentration | ≥5.0 mg/mL |
| För användning med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 3746 |
| Namn | Human KV3.1 (KCNC1) (aa 116-160) Control Fragment |
| Kvantitet | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | C230009H10Rik; EPM7; FLJ41162; FLJ42249; FLJ43491; KCNC1; Kcr2-1; Krc2 1; KShIIIB; KV3.1; KV4; MGC129855; NGK2; NGK2-KV4; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium channel, voltage gated Shaw-related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Shaw; Shaw-related voltage-gated potassium channel protein 1; voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4 |
| Vanligt namn | KV3.1 (KCNC1) |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering