missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human L3MBTL2 Partial ORF (NP_113676, 642 a.a. - 705 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083746-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180537

  • 4515.00 SEK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: RQGSKKPLLEDDPQGARKISSEPVPGEIIAVRVKEEHLDVASPDKASSPELPVSVENIKQETDD
Specifications

Specifications

NP_113676
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.78kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RQGSKKPLLEDDPQGARKISSEPVPGEIIAVRVKEEHLDVASPDKASSPELPVSVENIKQETDD
RUO
L3MBTL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
83746
L3MBTL2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
H-l(3)mbt-l/L3MBT
L3MBTL2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers