missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LCP2 Full-length ORF (AAH16618, 1 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003937-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117341

  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function. [provided by RefSeq]

Sequence: MALRNVPFRSEVLGWDPDSLADYFKKLNYKDCEKAVKKYHIDGARFLNLTENDIQKFPKLRVPILSKLSQEINKNEERRSIFTRKPQVPRFPEETESHEEDNGGWSSFEEDDYESPNDDQDGEDDGDYESPNEEEEAPVEDDADYEPPPSNDEEALQNSILPAKPFPNSNSMYIDRPPSGKTPQQPPVPPQRPMAALPPPPAGRNHSPLPPPQTNHEEPSRSRNHKTAKLPAPSIDRSTKPPLDRSLAPFDREPFTLGKKPPFSDKPSIPAGRSLGEHLPKIQKPPLPPTTERHERSSPLPGKKPPVPKHGWGPDRRENDEDDVHQRPLPQPALLPMSSNTFPSRSTKPSPMNPLPSSHMPGAFSESNSSFPQSASLPPYFSQGPSNRPPIRAEGRNFPLPLPNKPRPPSPAEEENSLNEEWYVSYITRPEAEAALRKINQDGTFLVRDSSKKTTTNPYVLMVLYKDKVYNIQIRYQKESQVYLLGTGLRGKEDFLSVSDIIDYFRKMPLLLIDGKNRGSRYQCTLTHAAGYP
Specifications

Specifications

AAH16618
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.37kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLP-76/SLP76
LCP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3937
LCP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALRNVPFRSEVLGWDPDSLADYFKKLNYKDCEKAVKKYHIDGARFLNLTENDIQKFPKLRVPILSKLSQEINKNEERRSIFTRKPQVPRFPEETESHEEDNGGWSSFEEDDYESPNDDQDGEDDGDYESPNEEEEAPVEDDADYEPPPSNDEEALQNSILPAKPFPNSNSMYIDRPPSGKTPQQPPVPPQRPMAALPPPPAGRNHSPLPPPQTNHEEPSRSRNHKTAKLPAPSIDRSTKPPLDRSLAPFDREPFTLGKKPPFSDKPSIPAGRSLGEHLPKIQKPPLPPTTERHERSSPLPGKKPPVPKHGWGPDRRENDEDDVHQRPLPQPALLPMSSNTFPSRSTKPSPMNPLPSSHMPGAFSESNSSFPQSASLPPYFSQGPSNRPPIRAEGRNFPLPLPNKPRPPSPAEEENSLNEEWYVSYITRPEAEAALRKINQDGTFLVRDSSKKTTTNPYVLMVLYKDKVYNIQIRYQKESQVYLLGTGLRGKEDFLSVSDIIDYFRKMPLLLIDGKNRGSRYQCTLTHAAGYP
RUO
LCP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers