missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGALS4 Partial ORF (NP_006140, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003960-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137381

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. [provided by RefSeq]

Sequence: VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFI
Specifications

Specifications

NP_006140
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFI
RUO
LGALS4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3960
LGALS4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAL4/L36LBP
LGALS4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers