missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LGR7 Partial ORF (NP_067647, 68 a.a. - 162 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00059350-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: WSMQFDKYFASYYKMTSQYPFEAETPECLVGSVPVQCLCQGLELDCDETNLRAVPSVSSNVTAMSLQWNLIRKLPPDCFKNYHDLQKLYLQNNKISpecifications
NP_067647 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WSMQFDKYFASYYKMTSQYPFEAETPECLVGSVPVQCLCQGLELDCDETNLRAVPSVSSNVTAMSLQWNLIRKLPPDCFKNYHDLQKLYLQNNKI | |
RUO | |
RXFP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
59350 | |
LGR7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LGR7/LGR7.1/LGR7.10/LGR7.2/MGC138347/MGC142177/RXFPR1 | |
RXFP1 | |
Recombinant | |
wheat germ expression system |