Learn More
Abnova™ Human LHFPL5 Full-length ORF (NP_872354.1, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00222662-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in this gene result in deafness in humans, and a mutation in a similar gene in mice results in deafness and vestibular dysfunction with severe degeneration of the organ of Corti. It is proposed to function in hair bundle morphogenesis. [provided by RefSeq]
Sequence: MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEVSpecifications
NP_872354.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB67/MGC33835/TMHS/dJ510O8.8 | |
LHFPL5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
222662 | |
LHFPL5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEV | |
RUO | |
LHFPL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |