missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LHX2 Full-length ORF (NP_004780.3, 1 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009355-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133562

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq]

Sequence: MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLF
Specifications

Specifications

NP_004780.3
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LH2/MGC138390/hLhx2
LHX2
Yes
wheat germ expression system
Liquid
9355
LHX2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLF
RUO
LHX2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates