missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human LIF ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP1-91186
This item is not returnable.
View return policy
Description
The Human LIF ELISA Kit (Colorimetric) measures human LIF in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
3.15 - 200pg/mL | |
ELISA Kit | |
The cross-reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human LIF positive serum. No cross-reactivity or interference was detected. | |
ELISA | |
LIF | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
LIF |
0.7pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
Human | |
3976 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonalantibody to human LIF, Biotin-Conjugate anti-human LIF monoclonal antibody, Streptavidin-HRP, Human LIF Standard, 400 pg/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic diffe |