Läs mer
Human LILRA1/CD85i/LIR-6 Antibody, R&D Systems™
Beskrivning
LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Immunocytochemistry.
Specifikationer
Specifikationer
| Antigen | LIR-6/LILRA1 |
| Användningsområden | Immunocytochemistry |
| Klassificering | Monoclonal |
| Klona | 586439 |
| Konjugera | Unconjugated |
| Utspädning | Immunocytochemistry 8-25 μ/mL |
| Formulering | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Genaccessionsnr. | O75019 |
| Gene Alias | CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563 |
| Värd art | Mouse |
| Visa mer |