missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LOC285382 Full-length ORF (NP_001020437.1, 1 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00285382-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16130014

  • 5030.41 SEK / 25µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTV
Specifications

Specifications

NP_001020437.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C3orf70
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
285382
LOC285382 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTV
RUO
C3orf70
Yes
Videos
SDS
Documents
Certificates