missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LOC493829 Partial ORF (NP_001008275.1, 378 a.a. - 477 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number NP_001008275.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 493829
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16133007
View Documents Promotion Details
Abnova™
H00493829-Q01.10UG
10 ug
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16143007
View Documents Promotion Details
Abnova™
H00493829-Q01.25UG
25 ug
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: QGLWLLGLREGKILEAHVEAKEPRALRSPERRPTRIGLYLSFGDGVLSFYDASDADALVPLFAFHERLPRPVYPFFDVCWHDKGKNAQPLLLVGPEGAEA
Specifications

Specifications

NP_001008275.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TRIM72
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
493829
LOC493829 (Human) Recombinant Protein (Q01)
QGLWLLGLREGKILEAHVEAKEPRALRSPERRPTRIGLYLSFGDGVLSFYDASDADALVPLFAFHERLPRPVYPFFDVCWHDKGKNAQPLLLVGPEGAEA
RUO
TRIM72
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications